Assay Details
Gene Symbol: LOC5575302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853855.1 | XP_021709547.1 | 4 - 5 | 1669 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5575302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021853855.1 | XP_021709547.1 | 4 - 5 | 1669 | 56 |
Set Membership: |
![]() ![]() ![]() |