Assay Details
Gene Symbol: LOC5569575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658592.3 | XP_001658642.1 | 3 - 4 | 1149 | 87 | |
XM_021844046.1 | XP_021699738.1 | 4 - 5 | 1222 | 87 | ||
GenBank mRNA | AY433176.1 | - | 3 - 4 | 1138 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
