Assay Details
Gene Symbol: LOC5565856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650147.2 | XP_001650197.1 | 2 - 3 | 381 | 147 | |
GenBank mRNA | AY433195.1 | - | 2 - 3 | 356 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
