Assay Details
Gene Symbol: LOC5570372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851132.1 | XP_021706824.1 | 3 - 4 | 1364 | 68 | |
XM_021851133.1 | XP_021706825.1 | 3 - 4 | 1246 | 68 | ||
XM_021851134.1 | XP_021706826.1 | 3 - 4 | 1181 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
