Assay Details
Gene Symbol: LOC5579843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854134.1 | XP_021709826.1 | 3 - 4 | 366 | 97 | |
XM_021854135.1 | XP_021709827.1 | 3 - 4 | 371 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
