Assay Details
Gene Symbol: LOC5563805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648039.2 | XP_001648089.1 | 4 - 5 | 895 | 67 | |
XM_021845603.1 | XP_021701295.1 | 4 - 5 | 916 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
