Assay Details
Gene Symbol: LOC5564484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648781.2 | XP_001648831.2 | 2 - 3 | 616 | 68 | |
GenBank mRNA | KF227904.1 | - | 2 - 3 | 622 | 68 | |
KF227905.1 | - | 1 - 2 | 353 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
