Assay Details
Gene Symbol: LOC5564484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648781.2 | XP_001648831.2 | 3 - 4 | 806 | 110 | |
GenBank mRNA | KF227904.1 | - | 3 - 4 | 812 | 110 | |
KF227905.1 | - | 2 - 3 | 543 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
