Assay Details
Gene Symbol: LOC5563807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648042.2 | XP_001648092.1 | 2 - 3 | 713 | 67 | |
XM_001648043.2 | XP_001648093.1 | 2 - 3 | 657 | 67 | ||
XM_021845605.1 | XP_021701297.1 | 2 - 3 | 583 | 67 | ||
XM_021845606.1 | XP_021701298.1 | 2 - 3 | 690 | 67 | ||
XM_021845607.1 | XP_021701299.1 | 2 - 3 | 646 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
