Assay Details
Gene Symbol: LOC5573854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654831.2 | XP_001654881.1 | 5 - 6 | 1160 | 70 | |
XM_021845591.1 | XP_021701283.1 | 4 - 5 | 1074 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
