Assay Details
Gene Symbol: LOC5566857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651234.2 | XP_001651284.1 | 1 - 2 | 196 | 135 | |
GenBank mRNA | AY433257.1 | - | 1 - 2 | 132 | 135 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001651234.2 transcripts.
Set Membership: |
![]() ![]() |
