Assay Details
Gene Symbol: LOC5566858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651231.2 | XP_001651281.2 | 1 - 2 | 656 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5566858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651231.2 | XP_001651281.2 | 1 - 2 | 656 | 69 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |