Assay Details
Gene Symbol: LOC5564770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649072.2 | XP_001649122.1 | 1 - 2 | 163 | 108 | |
XM_001649073.2 | XP_001649123.1 | 1 - 2 | 163 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001649072.2, XM_001649073.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
