Assay Details
Gene Symbol: LOC110673977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845454.1 | XP_021701146.1 | 3 - 4 | 473 | 82 | |
XM_021845455.1 | XP_021701147.1 | 3 - 4 | 470 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
