Assay Details
Gene Symbol: LOC23687815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494872.2 | XP_011493174.1 | 2 - 3 | 617 | 59 | |
XM_021849256.1 | XP_021704948.1 | 3 - 4 | 617 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
