Assay Details
Gene Symbol: LOC23687815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494872.2 | XP_011493174.1 | 4 - 5 | 1019 | 62 | |
XM_021849256.1 | XP_021704948.1 | 5 - 6 | 1019 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
