Assay Details
Gene Symbol: LOC5576380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662546.3 | XP_001662596.2 | 3 - 4 | 420 | 108 | |
XM_021844028.1 | XP_021699720.1 | 3 - 4 | 462 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001662546.3, XM_021844028.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
