Assay Details
Gene Symbol: LOC5576380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662546.3 | XP_001662596.2 | 8 - 9 | 3622 | 80 | |
XM_021844028.1 | XP_021699720.1 | 8 - 9 | 3664 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
