Assay Details
Gene Symbol: LOC5569861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846293.1 | XP_021701985.1 | 2 - 3 | 1382 | 88 | |
XM_021846294.1 | XP_021701986.1 | 2 - 3 | 1987 | 88 | ||
XM_021846295.1 | XP_021701987.1 | 2 - 3 | 1621 | 88 | ||
XM_021846296.1 | XP_021701988.1 | 2 - 3 | 1424 | 88 | ||
XM_021846297.1 | XP_021701989.1 | 2 - 3 | 1294 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
