Assay Details
Gene Symbol: LOC5570412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843135.1 | XP_021698827.1 | 5 - 6 | 2668 | 102 | |
XM_021843137.1 | XP_021698829.1 | 5 - 6 | 1741 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
