Assay Details
Gene Symbol: LOC5575983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838777.1 | XP_021694469.1 | 2 - 3 | 2823 | 95 | |
XM_021838778.1 | XP_021694470.1 | 3 - 4 | 1450 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
