Assay Details
Gene Symbol: LOC5565307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649615.2 | XP_001649665.2 | 1 - 2 | 542 | 102 | |
XM_001649616.2 | XP_001649666.2 | 1 - 2 | 542 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001649615.2, XM_001649616.2 transcripts.
Set Membership: |
![]() ![]() |
