Assay Details
Gene Symbol: LOC5565307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649615.2 | XP_001649665.2 | 2 - 3 | 1160 | 71 | |
XM_001649616.2 | XP_001649666.2 | 2 - 3 | 1160 | 71 |
