Assay Details
Gene Symbol: LOC5565307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649615.2 | XP_001649665.2 | 4 - 5 | 6008 | 77 | |
XM_001649616.2 | XP_001649666.2 | 4 - 5 | 6008 | 77 |
