Assay Details
Gene Symbol: LOC5565307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649615.2 | XP_001649665.2 | 5 - 6 | 6170 | 68 | |
XM_001649616.2 | XP_001649666.2 | 5 - 6 | 6170 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
