Assay Details
Gene Symbol: LOC5574043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843075.1 | XP_021698767.1 | 8 - 9 | 2047 | 91 | |
XM_021843076.1 | XP_021698768.1 | 8 - 9 | 1644 | 91 | ||
XM_021843078.1 | XP_021698770.1 | 7 - 8 | 2064 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
