Assay Details
Gene Symbol: LOC5574043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843075.1 | XP_021698767.1 | 2 - 3 | 775 | 105 | |
XM_021843076.1 | XP_021698768.1 | 2 - 3 | 372 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021843075.1, XM_021843076.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
