Assay Details
Gene Symbol: LOC5566381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846700.1 | XP_021702392.1 | 6 - 7 | 4191 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5566381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846700.1 | XP_021702392.1 | 6 - 7 | 4191 | 95 |
Set Membership: |
![]() ![]() ![]() |