Assay Details
Gene Symbol: LOC5574336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661320.2 | XP_001661370.1 | 2 - 3 | 917 | 67 | |
XM_021843224.1 | XP_021698916.1 | 2 - 3 | 917 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
