Assay Details
Gene Symbol: LOC5578317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845847.1 | XP_021701539.1 | 5 - 6 | 3021 | 68 | |
XM_021845848.1 | XP_021701540.1 | 5 - 6 | 2407 | 68 | ||
XM_021845849.1 | XP_021701541.1 | 5 - 6 | 2353 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
