Assay Details
Gene Symbol: LOC5575871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655823.2 | XP_001655873.1 | 1 - 2 | 376 | 84 | |
XM_021849172.1 | XP_021704864.1 | 1 - 2 | 376 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001655823.2, XM_021849172.1 transcripts.
Set Membership: |
![]() ![]() |
