Assay Details
Gene Symbol: LOC5567858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651845.2 | XP_001651895.1 | 7 - 8 | 2338 | 89 | |
XM_021849417.1 | XP_021705109.1 | 7 - 8 | 2042 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
