Assay Details
Gene Symbol: LOC5570306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854285.1 | XP_021709977.1 | 4 - 5 | 1586 | 134 | |
XM_021854286.1 | XP_021709978.1 | 3 - 4 | 1512 | 134 | ||
XM_021854287.1 | XP_021709979.1 | 3 - 4 | 1643 | 134 | ||
XM_021854288.1 | XP_021709980.1 | 3 - 4 | 1452 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
