Assay Details
Gene Symbol: LOC5570306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854285.1 | XP_021709977.1 | 3 - 4 | 686 | 115 | |
XM_021854286.1 | XP_021709978.1 | 2 - 3 | 612 | 115 | ||
XM_021854287.1 | XP_021709979.1 | 2 - 3 | 743 | 115 | ||
XM_021854288.1 | XP_021709980.1 | 2 - 3 | 552 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021854285.1, XM_021854286.1, XM_021854287.1, XM_021854288.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
