Assay Details
Gene Symbol: LOC5566597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850317.1 | XP_021706009.1 | 4 - 5 | 1564 | 63 | |
XM_021850318.1 | XP_021706010.1 | 4 - 5 | 1333 | 63 | ||
XM_021850319.1 | XP_021706011.1 | 4 - 5 | 1310 | 63 | ||
XM_021850321.1 | XP_021706013.1 | 3 - 4 | 1120 | 63 | ||
XM_021850322.1 | XP_021706014.1 | 3 - 4 | 1109 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
