Assay Details
Gene Symbol: LOC5565598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649896.2 | XP_001649946.1 | 3 - 4 | 1401 | 67 | |
XM_021838095.1 | XP_021693787.1 | 3 - 4 | 1401 | 67 | ||
XM_021838096.1 | XP_021693788.1 | 3 - 4 | 1358 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
