Assay Details
Gene Symbol: LOC5575865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655820.3 | XP_001655870.1 | 3 - 4 | 751 | 108 | |
XM_021849155.1 | XP_021704847.1 | 4 - 5 | 719 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
