Assay Details
Gene Symbol: LOC5570324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653101.2 | XP_001653151.2 | 1 - 2 | 948 | 64 | |
XM_021843821.1 | XP_021699513.1 | 1 - 2 | 950 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
