Assay Details
Gene Symbol: LOC5564047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648307.2 | XP_001648357.1 | 1 - 2 | 128 | 67 | |
GenBank mRNA | AY431666.1 | - | 1 - 2 | 44 | 67 | |
AY432729.1 | - | 1 - 2 | 44 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001648307.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
