Assay Details
Gene Symbol: LOC5563904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648169.2 | XP_001648219.1 | 1 - 2 | 99 | ||
GenBank mRNA | AY431744.1 | - | 1 - 2 | 155 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
