Assay Details
Gene Symbol: LOC5565257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649566.2 | XP_001649616.2 | 6 - 7 | 2808 | 88 | |
XM_021850000.1 | XP_021705692.1 | 9 - 10 | 4758 | 88 | ||
XM_021850006.1 | XP_021705698.1 | 7 - 8 | 2766 | 88 | ||
XM_021850014.1 | XP_021705706.1 | 8 - 9 | 3115 | 88 | ||
XM_021850019.1 | XP_021705711.1 | 8 - 9 | 3118 | 88 | ||
XM_021850021.1 | XP_021705713.1 | 8 - 9 | 3098 | 88 | ||
XM_021850026.1 | XP_021705718.1 | 8 - 9 | 3101 | 88 | ||
XM_021850034.1 | XP_021705726.1 | 7 - 8 | 2913 | 88 | ||
GenBank mRNA | AY431790.1 | - | 1 - 2 | 78 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
