Assay Details
Gene Symbol: LOC5565257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649566.2 | XP_001649616.2 | 7 - 8 | 3260 | 63 | |
XM_021850000.1 | XP_021705692.1 | 10 - 11 | 5210 | 63 | ||
XM_021850006.1 | XP_021705698.1 | 8 - 9 | 3218 | 63 | ||
XM_021850014.1 | XP_021705706.1 | 9 - 10 | 3567 | 63 | ||
XM_021850019.1 | XP_021705711.1 | 9 - 10 | 3570 | 63 | ||
XM_021850021.1 | XP_021705713.1 | 9 - 10 | 3550 | 63 | ||
XM_021850026.1 | XP_021705718.1 | 9 - 10 | 3553 | 63 | ||
XM_021850034.1 | XP_021705726.1 | 8 - 9 | 3365 | 63 | ||
GenBank mRNA | AY431790.1 | - | 2 - 3 | 530 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
