Assay Details
Gene Symbol: LOC5571150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653416.2 | XP_001653466.1 | 3 - 4 | 479 | 85 | |
GenBank mRNA | AY431967.1 | - | 3 - 4 | 389 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
