Assay Details
Gene Symbol: LOC5565500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849454.1 | XP_021705146.1 | 3 | 2809 | 63 | |
GenBank mRNA | AY432035.1 | - | 1 | 219 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
