Assay Details
Gene Symbol: LOC5577779 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663494.2 | XP_001663544.1 | 5 - 6 | 771 | 87 | |
XM_001663495.2 | XP_001663545.1 | 5 - 6 | 771 | 87 | ||
XM_021854632.1 | XP_021710324.1 | 5 - 6 | 771 | 87 | ||
XM_021854633.1 | XP_021710325.1 | 5 - 6 | 771 | 87 | ||
XM_021854634.1 | XP_021710326.1 | 5 - 6 | 765 | 87 | ||
XM_021854635.1 | XP_021710327.1 | 5 - 6 | 771 | 87 | ||
XM_021854636.1 | XP_021710328.1 | 5 - 6 | 771 | 87 | ||
XM_021854637.1 | XP_021710329.1 | 6 - 7 | 986 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
