Assay Details
Gene Symbol: LOC5566303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855224.1 | XP_021710916.1 | 3 - 4 | 1020 | 73 | |
XM_021855225.1 | XP_021710917.1 | 3 - 4 | 1020 | 73 |
