Assay Details
Gene Symbol: LOC5574332 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838939.1 | XP_021694631.1 | 8 | 3560 | 87 | |
XM_021838940.1 | XP_021694632.1 | 8 | 3546 | 87 | ||
XM_021838941.1 | XP_021694633.1 | 8 | 3526 | 87 | ||
XM_021838942.1 | XP_021694634.1 | 8 | 3533 | 87 | ||
XM_021838943.1 | XP_021694635.1 | 8 | 3522 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
