Assay Details
Gene Symbol: LOC5571574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845692.1 | XP_021701384.1 | 4 - 5 | 2380 | 92 | |
XM_021845698.1 | XP_021701390.1 | 4 - 5 | 2380 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
