Assay Details
Gene Symbol: LOC5573080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846435.1 | XP_021702127.1 | 5 - 6 | 906 | 120 | |
XM_021846436.1 | XP_021702128.1 | 5 - 6 | 851 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
