Assay Details
Gene Symbol: LOC23687777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854883.1 | XP_021710575.1 | 2 - 3 | 6166 | 77 | |
XM_021854884.1 | XP_021710576.1 | 3 - 4 | 5969 | 77 | ||
XM_021854885.1 | XP_021710577.1 | 3 - 4 | 5974 | 77 | ||
XM_021854886.1 | XP_021710578.1 | 2 - 3 | 5355 | 77 | ||
XM_021854887.1 | XP_021710579.1 | 2 - 3 | 5420 | 77 |
